Learn More
MSC Vestil Beam Tongs, 6000 Lb., 10 In. Max Width, ASME B30.20
Catalog No. 5000533467
Encompass
These heavy-duty beam tongs are engineered to efficiently lift I-beams using overhead lifting devices. Their scissor-style mechanism leverages the load's weight to provide an increasingly stable and secure grip. Proper centering and guidance of loads are essential during all lifting operations.
  • Designed for lifting I-beams.
  • Scissor-style mechanism provides a stable grip that strengthens with heavier loads.
  • Includes a lifting ring for easy use with an overhead hoist.
  • Manufactured to ASME B30.20 standards.
  • Offers a maximum beam width of 10 in.
  • Features a 6,000 lb. uniform capacity.
  • Constructed from steel with a tough baked-in powder-coated yellow finish.
Catalog No. 50-005-33467 Supplier MSC Supplier No. 04754560
These heavy-duty beam tongs are engineered to efficiently lift I-beams using overhead lifting devices. Their scissor-style mechanism leverages the load's weight to provide an increasingly stable and secure grip. Proper centering and guidance of loads are essential during all lifting operations.
  • Designed for lifting I-beams.
  • Scissor-style mechanism provides a stable grip that strengthens with heavier loads.
  • Includes a lifting ring for easy use with an overhead hoist.
  • Manufactured to ASME B30.20 standards.
  • Offers a maximum beam width of 10 in.
  • Features a 6,000 lb. uniform capacity.
  • Constructed from steel with a tough baked-in powder-coated yellow finish.