Learn More
MSC Vestil Caster, Polyurethane, 6 x 2 in, 1232 lb, Rigid
Catalog No. 5000529392
Encompass
This polyurethane caster is designed for a wide range of environments, offering excellent resistance to various chemicals. It ensures an easy roll for ergonomic applications and provides good floor preservation. Suitable for both indoor and outdoor settings, this caster combines durability with efficient performance.
  • Non-marking
  • Lightweight rust resistant aluminum core
  • Hardened bearing and sealed raceway
  • Chemical and moisture resistant
  • Anti-corrosive and rust resistant zinc plated finish
  • Grease zerk fittings
  • Ball bearings
  • Roller bearings
Catalog No. 50-005-29392 Supplier MSC Supplier No. 17137324
This polyurethane caster is designed for a wide range of environments, offering excellent resistance to various chemicals. It ensures an easy roll for ergonomic applications and provides good floor preservation. Suitable for both indoor and outdoor settings, this caster combines durability with efficient performance.
  • Non-marking
  • Lightweight rust resistant aluminum core
  • Hardened bearing and sealed raceway
  • Chemical and moisture resistant
  • Anti-corrosive and rust resistant zinc plated finish
  • Grease zerk fittings
  • Ball bearings
  • Roller bearings