Learn More
MSC Vestil Drum Cover, Polyethylene, Closed Head, 55 Gallon, 25 in
Catalog No. 5000534989
Encompass
This round polyethylene drum cover is designed to protect 55-gallon closed steel head drums from bung seepage and outdoor storage elements. Constructed from flexible, thick polyethylene with an ultraviolet screen, it offers reliable use and protection.
  • Constructed from flexible, thick polyethylene.
  • Features an ultraviolet screen for enhanced durability.
  • Designed to protect drums from bung seepage.
  • Suitable for unprotected outdoor storage situations.
  • Provides years of reliable use.
  • Ideal for 55-gallon closed steel head drums.
Catalog No. 50-005-34989 Supplier MSC Supplier No. 62023536
This round polyethylene drum cover is designed to protect 55-gallon closed steel head drums from bung seepage and outdoor storage elements. Constructed from flexible, thick polyethylene with an ultraviolet screen, it offers reliable use and protection.
  • Constructed from flexible, thick polyethylene.
  • Features an ultraviolet screen for enhanced durability.
  • Designed to protect drums from bung seepage.
  • Suitable for unprotected outdoor storage situations.
  • Provides years of reliable use.
  • Ideal for 55-gallon closed steel head drums.