Learn More
MSC Vestil Drum Sling, Galvanized Steel, 2000 lb, 30-55 Gallon Drums
Catalog No. 5000533953
Encompass
This drum sling is designed for quickly and easily lifting 30- and 55-gallon steel drums in a horizontal position. Made from galvanized steel, it has a 2000 lb uniform capacity and can handle both open and closed head drums. Its large, heavy-duty steel lifting ring ensures compatibility with most overhead lifting systems.
  • Ideal for lifting 30- and 55-gallon drums quickly and easily
  • Handles open- or closed-head steel drums
  • Compatible with most overhead lifting systems
  • Features a 2000 lb uniform capacity
  • Constructed from galvanized steel for durability
Catalog No. 50-005-33953 Supplier MSC Supplier No. 30123608
This drum sling is designed for quickly and easily lifting 30- and 55-gallon steel drums in a horizontal position. Made from galvanized steel, it has a 2000 lb uniform capacity and can handle both open and closed head drums. Its large, heavy-duty steel lifting ring ensures compatibility with most overhead lifting systems.
  • Ideal for lifting 30- and 55-gallon drums quickly and easily
  • Handles open- or closed-head steel drums
  • Compatible with most overhead lifting systems
  • Features a 2000 lb uniform capacity
  • Constructed from galvanized steel for durability