Learn More
MSC Vestil Floor Lock, Steel, 8 in Height, 6 in Caster
Catalog No. 5000510213
Encompass
Floor locks can be used on platform trucks, fixture dollies, or equipment with casters. Simple foot pressure immobilizes the equipment. It locks when a large friction face is pressed against the floor, and to release, simply kick the release rod. The brake is independent of casters and can be installed at any convenient location on the cart. It features a vertical mounting plate designed to mount to the sides of carts and other portable products. The spring-loaded pad accommodates uneven floors, and units mount with the pad 1 inch off the floor when retracted.
  • Lock constricts movement of unit.
  • Attachable to moving platform carts.
  • Simple locking and unlocking function.
  • Able to be used on rough surfaces.
Catalog No. 50-005-10213 Supplier MSC Supplier No. 99676363
Floor locks can be used on platform trucks, fixture dollies, or equipment with casters. Simple foot pressure immobilizes the equipment. It locks when a large friction face is pressed against the floor, and to release, simply kick the release rod. The brake is independent of casters and can be installed at any convenient location on the cart. It features a vertical mounting plate designed to mount to the sides of carts and other portable products. The spring-loaded pad accommodates uneven floors, and units mount with the pad 1 inch off the floor when retracted.
  • Lock constricts movement of unit.
  • Attachable to moving platform carts.
  • Simple locking and unlocking function.
  • Able to be used on rough surfaces.