Learn More
MSC Vestil Forklift Boom, 4000 lb, 7-1/4 x 2-1/2 in Fork Pocket
Catalog No. 5000511110
Encompass
Convert your fork truck into a hook with an anchor in a matter of seconds. This easy-to-attach hoisting hook does not require special tools. It is secured to the fork truck by means of a long safety restraint and screw clamps, and includes a zinc plated hook with shackle.
  • Heavy-duty solid steel construction provides long lasting rugged use
  • Double-fork style securely slides onto both fork truck forks
  • Safety chain with snap hook attaches unit to carriage of fork truck
  • Forged steel hook with safety latch ensures worker safety
Catalog No. 50-005-11110 Supplier MSC Supplier No. 04754651
Convert your fork truck into a hook with an anchor in a matter of seconds. This easy-to-attach hoisting hook does not require special tools. It is secured to the fork truck by means of a long safety restraint and screw clamps, and includes a zinc plated hook with shackle.
  • Heavy-duty solid steel construction provides long lasting rugged use
  • Double-fork style securely slides onto both fork truck forks
  • Safety chain with snap hook attaches unit to carriage of fork truck
  • Forged steel hook with safety latch ensures worker safety