Learn More
MSC Vestil Forklift Fork Extensions, 4000 lb Capacity, 4 x 2 inch Fork Pocket, Steel
Catalog No. 5000528306
Encompass
These Forklift Fork Extensions are designed to provide additional support for lifting long or large objects with a fork truck. They are constructed from all-steel and feature a steel retaining strap to prevent them from sliding off the forks during use. Designed for use with Class 2 and Class 3 forks, these extensions ensure enhanced safety and efficiency in material handling operations.
  • Provides extra support for lifting long or large objects.
  • Features a steel retaining strap to prevent sliding.
  • Designed for use with Class 2 and Class 3 forks only.
  • Constructed from all-steel for durability.
  • Includes warning and operating stickers for safety.
  • Finished with a powder coat safety yellow for visibility.
  • OSHA regulations state extensions should be no more than 150% of existing fork length.
Catalog No. 50-005-28306 Supplier MSC Supplier No. 30116024
These Forklift Fork Extensions are designed to provide additional support for lifting long or large objects with a fork truck. They are constructed from all-steel and feature a steel retaining strap to prevent them from sliding off the forks during use. Designed for use with Class 2 and Class 3 forks, these extensions ensure enhanced safety and efficiency in material handling operations.
  • Provides extra support for lifting long or large objects.
  • Features a steel retaining strap to prevent sliding.
  • Designed for use with Class 2 and Class 3 forks only.
  • Constructed from all-steel for durability.
  • Includes warning and operating stickers for safety.
  • Finished with a powder coat safety yellow for visibility.
  • OSHA regulations state extensions should be no more than 150% of existing fork length.