Learn More
MSC Vestil Gantry Sling Rack, Stationary 2000 lb, Portable 1200 lb, Steel, Blue
Catalog No. 5000528273
Encompass
The Gantry Sling Rack is a versatile storage solution designed for manufacturing and industrial facilities. It features strong steel construction with a baked-in powder-coated finish for enhanced durability and allows for the hanging of both large and small items. This sling rack can be configured as either a stationary or portable unit, with included casters, mounting pads, adjustable feet, and an anchor bolt kit to suit various operational needs.
  • Strong steel construction with a baked-in blue powder coat finish
  • Middle rack is adjustable
  • Eight double hooks are adjustable
  • Includes four adjustable feet with 1 in. of adjustability
  • Includes four 3 in. diameter stem casters with brakes
  • Includes an anchor bolt kit
  • Provides high visibility
  • Robust assembly for long-term use
  • Accessible from both sides
Catalog No. 50-005-28273 Supplier MSC Supplier No. 38258364
The Gantry Sling Rack is a versatile storage solution designed for manufacturing and industrial facilities. It features strong steel construction with a baked-in powder-coated finish for enhanced durability and allows for the hanging of both large and small items. This sling rack can be configured as either a stationary or portable unit, with included casters, mounting pads, adjustable feet, and an anchor bolt kit to suit various operational needs.
  • Strong steel construction with a baked-in blue powder coat finish
  • Middle rack is adjustable
  • Eight double hooks are adjustable
  • Includes four adjustable feet with 1 in. of adjustability
  • Includes four 3 in. diameter stem casters with brakes
  • Includes an anchor bolt kit
  • Provides high visibility
  • Robust assembly for long-term use
  • Accessible from both sides