Learn More
MSC Vestil Gas Cylinder Wall Bracket, 2 Cylinder, Plastic, 4-12 in Cylinder Fit
Catalog No. 5000531750
Encompass
This gas cylinder wall bracket is designed to securely hold two cylinders. Measuring 29-1/4 in x 3 in x 4 in, it accommodates cylinders with diameters ranging from 4-12 inches. Constructed from molded plastic, it provides solid and secure use with a lightweight design for easy handling.
  • Wall mounted design
  • Easy to install
  • Cylinder strap secures cylinder to bracket
  • Accommodates cylinders from 4 to 12 in diameter
  • Made from durable plastic
  • Yellow color for high visibility
Catalog No. 50-005-31750 Supplier MSC Supplier No. 62050349
This gas cylinder wall bracket is designed to securely hold two cylinders. Measuring 29-1/4 in x 3 in x 4 in, it accommodates cylinders with diameters ranging from 4-12 inches. Constructed from molded plastic, it provides solid and secure use with a lightweight design for easy handling.
  • Wall mounted design
  • Easy to install
  • Cylinder strap secures cylinder to bracket
  • Accommodates cylinders from 4 to 12 in diameter
  • Made from durable plastic
  • Yellow color for high visibility