Learn More
MSC Vestil Gas Cylinder Wall Bracket, 36 in Long, Steel, Holds 3 Cylinders
Catalog No. 5000534925
Encompass
This wall-mounted cylinder bracket offers a simple, easy-to-install design for securing gas cylinders. It features a strap to hold cylinders firmly in place and includes rubber guards on the edges for added protection. Please note that installation hardware is not included.
  • Simple design is easy to install.
  • Cylinder strap secures cylinder to bracket.
  • Solid construction offers safe and secure use.
  • Lightweight design is easy to handle.
Catalog No. 50-005-34925 Supplier MSC Supplier No. 62050364
This wall-mounted cylinder bracket offers a simple, easy-to-install design for securing gas cylinders. It features a strap to hold cylinders firmly in place and includes rubber guards on the edges for added protection. Please note that installation hardware is not included.
  • Simple design is easy to install.
  • Cylinder strap secures cylinder to bracket.
  • Solid construction offers safe and secure use.
  • Lightweight design is easy to handle.