Learn More
MSC Vestil Heavy-Duty Roller Support, 1,760 Lb Capacity, 1 Roller, 14 in Roller Width
Catalog No. 5000527743
Encompass
This deluxe roller support is designed to provide extra assistance when working with long materials. It features a single chrome-plated roller and a heavy-duty cast-steel base for stability. The height can be easily adjusted with a friction lock screw or a gas charged cylinder, making it suitable for use in various machine and wood shop environments.
  • Easy height adjustment for different leveled machines
  • Lightweight construction for easy portability
  • Handy design acts as an extra hand when working with long materials
  • Cast steel base provides a solid and reliable foundation
Catalog No. 50-005-27743 Supplier MSC Supplier No. 38440731
This deluxe roller support is designed to provide extra assistance when working with long materials. It features a single chrome-plated roller and a heavy-duty cast-steel base for stability. The height can be easily adjusted with a friction lock screw or a gas charged cylinder, making it suitable for use in various machine and wood shop environments.
  • Easy height adjustment for different leveled machines
  • Lightweight construction for easy portability
  • Handy design acts as an extra hand when working with long materials
  • Cast steel base provides a solid and reliable foundation