Learn More
MSC Vestil Hose & Cable Ramp, Aluminum, 48 in Length, 10,000 lb Capacity
Catalog No. 5000533723
Encompass
This extruded aluminum hose and cable crossover ramp is designed to protect cables and hoses from damage by pedestrian and rolling traffic. It features heavy-duty aluminum construction for durability and an attractive appearance, with a single-channel design and serrated edges for maximum traction.
  • Protects cables and hoses from pedestrian and rolling traffic.
  • Heavy-duty aluminum extrusion construction ensures durability and an attractive appearance.
  • Features a single-channel design.
  • Serrated edges provide maximum traction.
  • Corrosion-resistant and long-lasting aluminum material.
  • Offers a uniform capacity of 10,000 lb for a single wheel.
Catalog No. 50-005-33723 Supplier MSC Supplier No. 30114425
This extruded aluminum hose and cable crossover ramp is designed to protect cables and hoses from damage by pedestrian and rolling traffic. It features heavy-duty aluminum construction for durability and an attractive appearance, with a single-channel design and serrated edges for maximum traction.
  • Protects cables and hoses from pedestrian and rolling traffic.
  • Heavy-duty aluminum extrusion construction ensures durability and an attractive appearance.
  • Features a single-channel design.
  • Serrated edges provide maximum traction.
  • Corrosion-resistant and long-lasting aluminum material.
  • Offers a uniform capacity of 10,000 lb for a single wheel.