Learn More
MSC Vestil Pail Lid, Steel, Standard Style, 5 Gallon
Catalog No. 5000536732
Encompass
This steel lid is designed for use with steel pails, offering a standard style. It securely seals 5 gallon open head steel pails and features lugs and a flow-in synthetic rubber gasket for a secure closure.
  • Designed to securely seal 5 gallon open head steel pails
  • Steel lid with lugs and a flow-in synthetic rubber gasket
  • Unique construction allows use in hot or cold applications
  • Meets UN 1A1/Y1 5/200 requirements
Catalog No. 50-005-36732 Supplier MSC Supplier No. 94558368
This steel lid is designed for use with steel pails, offering a standard style. It securely seals 5 gallon open head steel pails and features lugs and a flow-in synthetic rubber gasket for a secure closure.
  • Designed to securely seal 5 gallon open head steel pails
  • Steel lid with lugs and a flow-in synthetic rubber gasket
  • Unique construction allows use in hot or cold applications
  • Meets UN 1A1/Y1 5/200 requirements