Learn More
MSC Vestil Pipe Grab, Steel, 1,000 Lb, 5.5-7.0 in Pipe O.D.
Catalog No. 5000529250
Encompass
This heavy-duty pipe grab is designed for safely lifting cast iron pipe with an overhead lifting device. Constructed from durable steel, it features automatic operation for ease of use and a 1,000 Lb capacity. It is suitable for pipes with an outside diameter ranging from 5.5 to 7.0 inches.
  • Designed for lifting cast iron pipe with an overhead lifting device.
  • Automatic operation for easy use.
  • Heavy-duty steel construction for strength and durability.
  • Durable baked-in powder coated yellow finish.
  • For 5.5 to 7.0 in O.D. cast iron pipe or tube.
Catalog No. 50-005-29250 Supplier MSC Supplier No. 04755054
This heavy-duty pipe grab is designed for safely lifting cast iron pipe with an overhead lifting device. Constructed from durable steel, it features automatic operation for ease of use and a 1,000 Lb capacity. It is suitable for pipes with an outside diameter ranging from 5.5 to 7.0 inches.
  • Designed for lifting cast iron pipe with an overhead lifting device.
  • Automatic operation for easy use.
  • Heavy-duty steel construction for strength and durability.
  • Durable baked-in powder coated yellow finish.
  • For 5.5 to 7.0 in O.D. cast iron pipe or tube.