Learn More
MSC Vestil Plastic Face Molded Rubber Bumper, 4 Inch Projection, Fiber Reinforced
Catalog No. 5000528588
Encompass
This molded bumper features two-piece construction and is manufactured from fiber-reinforced prime rubber containing nylon and polyester, designed to withstand years of abusive pounding. It comes fully assembled and ready for installation on loading docks to protect trailers and the dock itself. Installation hardware is sold separately.
  • Plastic face molded rubber bumper.
  • Constructed from fiber reinforced prime rubber with nylon and polyester.
  • Dimensions: 11-13/16 in. x 12-13/16 in. x 3-7/8 in. (height x width x depth).
  • 4 inch projection.
  • Pre-drilled countersunk mounting holes for easy installation.
  • Accepts 3/4 in. anchor bolt diameter.
  • Net weight: 29 lbs.
  • 30 days limited warranty.
Catalog No. 50-005-28588 Supplier MSC Supplier No. 30114607
This molded bumper features two-piece construction and is manufactured from fiber-reinforced prime rubber containing nylon and polyester, designed to withstand years of abusive pounding. It comes fully assembled and ready for installation on loading docks to protect trailers and the dock itself. Installation hardware is sold separately.
  • Plastic face molded rubber bumper.
  • Constructed from fiber reinforced prime rubber with nylon and polyester.
  • Dimensions: 11-13/16 in. x 12-13/16 in. x 3-7/8 in. (height x width x depth).
  • 4 inch projection.
  • Pre-drilled countersunk mounting holes for easy installation.
  • Accepts 3/4 in. anchor bolt diameter.
  • Net weight: 29 lbs.
  • 30 days limited warranty.