Learn More
MSC Vestil Pry Bar Lever Dolly, 5,000 lb Load Limit, 5 ft Length, Steel
Catalog No. 5000513917
Encompass
The Prylever Bar is designed for dock workers, riggers, and freight handlers, enabling users to get under heavy objects for precise positioning. Constructed of steel with a powder coat finish, this unit offers long-lasting life and durability.
  • Prylever bar design provides individuals with the needed leverage.
  • Thick steel nose plate with a beveled edge allows users to easily get under loads.
  • Poly-on-steel wheels offer a strong, durable base.
  • Bar length gives the user an ergonomic edge to avoid injury.
Catalog No. 50-005-13917 Supplier MSC Supplier No. 64111990
The Prylever Bar is designed for dock workers, riggers, and freight handlers, enabling users to get under heavy objects for precise positioning. Constructed of steel with a powder coat finish, this unit offers long-lasting life and durability.
  • Prylever bar design provides individuals with the needed leverage.
  • Thick steel nose plate with a beveled edge allows users to easily get under loads.
  • Poly-on-steel wheels offer a strong, durable base.
  • Bar length gives the user an ergonomic edge to avoid injury.