Learn More
MSC Vestil Standard Casters, Phenolic Wheel, 6 inch Wheel Diameter, Roller Bearing
Catalog No. 5000513484
Encompass
These standard casters are designed for commercial and industrial applications, featuring a durable phenolic wheel and a robust roller bearing for smooth operation. With a 6 inch wheel diameter and a 1200 lb load capacity, they are suitable for a variety of heavy-duty tasks, offering reliable mobility and finished with a protective zinc coating.
  • Phenolic wheel material for durability.
  • Roller bearing type ensures smooth movement.
  • 6 inch wheel diameter for enhanced mobility.
  • 1200 lb load capacity for heavy-duty applications.
  • Designed for commercial and industrial use.
  • Zinc finish provides protection.
  • Mounts with holes for secure attachment.
Catalog No. 50-005-13484 Supplier MSC Supplier No. 17139122
These standard casters are designed for commercial and industrial applications, featuring a durable phenolic wheel and a robust roller bearing for smooth operation. With a 6 inch wheel diameter and a 1200 lb load capacity, they are suitable for a variety of heavy-duty tasks, offering reliable mobility and finished with a protective zinc coating.
  • Phenolic wheel material for durability.
  • Roller bearing type ensures smooth movement.
  • 6 inch wheel diameter for enhanced mobility.
  • 1200 lb load capacity for heavy-duty applications.
  • Designed for commercial and industrial use.
  • Zinc finish provides protection.
  • Mounts with holes for secure attachment.