Learn More
MSC Vestil Steel Rolling Ladder, 4 Step, 350 lb Capacity, Folding Design
Catalog No. 5000530718
Encompass
This folding ladder with wheels features a locking foldable design for space-saving storage and easy portability. Constructed from steel, it includes perforated steps for a non-slip surface and complies with ANSI 14.7 and OSHA 1910.29 specifications.
  • Locking foldable design saves on storage space when not in use
  • Folds after unlocking a spring-loaded detent pin
  • Tilts back on wheels for easy portability once folded
  • Made of steel, available in carbon steel or stainless steel options
  • Perforated steps provide a non-slip surface
  • Complies with ANSI 14.7 and OSHA 1910.29 specifications
Catalog No. 50-005-30718 Supplier MSC Supplier No. 89106033
This folding ladder with wheels features a locking foldable design for space-saving storage and easy portability. Constructed from steel, it includes perforated steps for a non-slip surface and complies with ANSI 14.7 and OSHA 1910.29 specifications.
  • Locking foldable design saves on storage space when not in use
  • Folds after unlocking a spring-loaded detent pin
  • Tilts back on wheels for easy portability once folded
  • Made of steel, available in carbon steel or stainless steel options
  • Perforated steps provide a non-slip surface
  • Complies with ANSI 14.7 and OSHA 1910.29 specifications