Learn More
MSC Vestil Vertical Positive Locking Plate Clamp, 4000 Lb Capacity, 0 to 1.19 in Grip
Catalog No. 5000513250
Encompass
This vertical positive locking plate clamp is designed for securely gripping and lifting single, clean, oil-free plates in a vertical position. Constructed from heavy-duty steel with a blue powder-coated finish, it features a serrated gripper made of hardened tool steel, and a forged head and gear. A locking mechanism ensures maximum safety, and the clamp meets ASME B30.20 specifications, offering a 2 to 1 safety factor for reliable, long-lasting use.
  • Heavy-duty steel construction for reliable, long-lasting use.
  • Unique design allows lifting of plate material in a vertical position.
  • Locking mechanism provides maximum safety.
  • Designed to meet ASME B30.20 standards.
  • Grips sheets securely until the release ring is pulled.
  • Serrated gripper made of hardened tool steel.
  • Forged head and gear components.
  • Safety factor of 2 to 1.
  • Features a powder coated finish.
Catalog No. 50-005-13250 Supplier MSC Supplier No. 04755013
This vertical positive locking plate clamp is designed for securely gripping and lifting single, clean, oil-free plates in a vertical position. Constructed from heavy-duty steel with a blue powder-coated finish, it features a serrated gripper made of hardened tool steel, and a forged head and gear. A locking mechanism ensures maximum safety, and the clamp meets ASME B30.20 specifications, offering a 2 to 1 safety factor for reliable, long-lasting use.
  • Heavy-duty steel construction for reliable, long-lasting use.
  • Unique design allows lifting of plate material in a vertical position.
  • Locking mechanism provides maximum safety.
  • Designed to meet ASME B30.20 standards.
  • Grips sheets securely until the release ring is pulled.
  • Serrated gripper made of hardened tool steel.
  • Forged head and gear components.
  • Safety factor of 2 to 1.
  • Features a powder coated finish.