Learn More
MSC Vestil Welding Cylinder Torch Cart, 500 lb Capacity, 16 in Foam Filled Wheels
Catalog No. 5000527360
Encompass
This welding cylinder torch cart is designed for transporting cylinders and cutting torches. It includes two tank compartments with heavy-duty ratchet straps and a convenient open-top storage tray. Steel brackets are included for holding cutting torch and hose. It features sturdy handles and large wheels for tilt-n-go portability and has a welded steel construction with a powder coat finish. This cart is not designed for use as a storage means for gas cylinders.
  • Heavy-duty ratchet straps secure tanks
  • Lockable storage box for regulators
  • Steel brackets safely hold cutting torch and hoses
  • Open-top storage tray for additional storage
Catalog No. 50-005-27360 Supplier MSC Supplier No. 68216613
This welding cylinder torch cart is designed for transporting cylinders and cutting torches. It includes two tank compartments with heavy-duty ratchet straps and a convenient open-top storage tray. Steel brackets are included for holding cutting torch and hose. It features sturdy handles and large wheels for tilt-n-go portability and has a welded steel construction with a powder coat finish. This cart is not designed for use as a storage means for gas cylinders.
  • Heavy-duty ratchet straps secure tanks
  • Lockable storage box for regulators
  • Steel brackets safely hold cutting torch and hoses
  • Open-top storage tray for additional storage