Learn More
Med Vet International Vet-lite 6" x 70", Cast Material, J0758D, 10 Rolls
Catalog No. 5000582137
Encompass_Preferred
This product was a popular product that was discontinued a few years ago in the veterinary field. It is now available once again. Constructed of thermoplastic white mesh. When heated in hot water, it turns into a soft malleable material that can be shaped into any configuration. Hardens as it cools.Can be reheated and reshapedPartially used material may be retained for future useWaterproofGloves not requiredLong shelf life
Catalog No. 50-005-82137 Supplier Med Vet International Supplier No. JOR758D
This product was a popular product that was discontinued a few years ago in the veterinary field. It is now available once again. Constructed of thermoplastic white mesh. When heated in hot water, it turns into a soft malleable material that can be shaped into any configuration. Hardens as it cools.Can be reheated and reshapedPartially used material may be retained for future useWaterproofGloves not requiredLong shelf life