Learn More
Santa Cruz Biotechnology VETBOND TISSUE ADHESIVE 3ML
Catalog No. NC0846393
Encompass
Vetbond Tissue Adhesive is a critical care product for use in wound care. Supplied as a 3 ml bottle. Vetbond Tissue Adhesive is used to quickly and conveniently close minor wounds. Vetbond often eliminates the need for sutures or bandages. Creates a strong bond that polymerizes in seconds. Binds wound edges together to promote natural healing. Vetbond Tissue Adhesive is blue, making application easier to see. Ingredients: Vetbond Tissue Adhesive contains N-butyl cyanoacrylate (>98% by weight), Hydroquinone (<1 % by weight), and blue dye (.01% by weight).Store in a cool, dry place. Maintain product at room temperature during use. Product can be refrigerated or frozen for extended shelf life before opening. Store bottle upright on shelf in a cool, dry place (away from heat). After opening, do not return bottle to the refrigerator.
Catalog No. NC0846393 Supplier Santa Cruz Biotechnology Supplier No. SC361931
Vetbond Tissue Adhesive is a critical care product for use in wound care. Supplied as a 3 ml bottle. Vetbond Tissue Adhesive is used to quickly and conveniently close minor wounds. Vetbond often eliminates the need for sutures or bandages. Creates a strong bond that polymerizes in seconds. Binds wound edges together to promote natural healing. Vetbond Tissue Adhesive is blue, making application easier to see. Ingredients: Vetbond Tissue Adhesive contains N-butyl cyanoacrylate (>98% by weight), Hydroquinone (<1 % by weight), and blue dye (.01% by weight).Store in a cool, dry place. Maintain product at room temperature during use. Product can be refrigerated or frozen for extended shelf life before opening. Store bottle upright on shelf in a cool, dry place (away from heat). After opening, do not return bottle to the refrigerator.