Learn More
Andwin Scientific VETEQUIP CHARCOL CANISTER
SDP
Catalog No. NC0475149
Encompass_Preferred
The Activated Charcoal Filter VaporGuard aid in controlling and monitoring the exposure to anesthetic waste gases. This product adsorbs passively. It is proven effective in removing waste anesthetic gas pollution.
Catalog No. NC0475149 Supplier Andwin Scientific Supplier No. 89012608
The Activated Charcoal Filter VaporGuard aid in controlling and monitoring the exposure to anesthetic waste gases. This product adsorbs passively. It is proven effective in removing waste anesthetic gas pollution.