Learn More
Colonial Medical Supply Co Inc OxyVet O2 Concentrator, Pressure Swing Adsorption Technology, Built-in Low O2 Alarm, 22 in
SDP
Catalog No. NC2152917
Encompass
The VetEquip OxyVet O2 Concentrator is an innovative oxygen generation system ideal for veterinary anesthesia setups. Utilizing Pressure Swing Adsorption technology, it provides a consistent supply of oxygen while offering significant cost savings over traditional tanks. Its compact design, quiet operation, and compatibility with various anesthesia systems enhance flexibility and safety in laboratory and surgical environments.
This item is not returnable. View return policy
Catalog No. NC2152917 Supplier Colonial Medical Supply Co Inc Supplier No. 931614
The VetEquip OxyVet O2 Concentrator is an innovative oxygen generation system ideal for veterinary anesthesia setups. Utilizing Pressure Swing Adsorption technology, it provides a consistent supply of oxygen while offering significant cost savings over traditional tanks. Its compact design, quiet operation, and compatibility with various anesthesia systems enhance flexibility and safety in laboratory and surgical environments.