Learn More
Med Vet International Vetericyn Plus VF Antimicrobial Otic Solution, 3oz
Catalog No. 503045267
Encompass_Preferred
Vetericyn Plus VF Antimicrobial Otic Solution is specially formulated for flushing and cleaning wounds and removing foreign debris and irritants from the outer ear and ear canal. It contains synthesized hypochlorous, a known antimicrobial also produced by the animal during phagocytosis that can be used on all otic tissue without harming fibroblast cells. Formula is pH balanced and does not irritate inflamed ears so animals do not resist treatment Contains no steroids, antibiotics, or alcohol Using Vetericyn prior to other medications helps prep otic tissues, thus allowing medications and other treatments to work more effectively  
Catalog No. 50-304-5267 Supplier Med Vet International Supplier No. PHVETERICYNOTIC
Vetericyn Plus VF Antimicrobial Otic Solution is specially formulated for flushing and cleaning wounds and removing foreign debris and irritants from the outer ear and ear canal. It contains synthesized hypochlorous, a known antimicrobial also produced by the animal during phagocytosis that can be used on all otic tissue without harming fibroblast cells. Formula is pH balanced and does not irritate inflamed ears so animals do not resist treatment Contains no steroids, antibiotics, or alcohol Using Vetericyn prior to other medications helps prep otic tissues, thus allowing medications and other treatments to work more effectively  