Learn More
Med Vet International Miltex Vantage Veterinary Feline Spay Pack
Catalog No. 502831606
Encompass_Preferred
The Miltex Vantage V96820 feline spay pack contains 19 mid-grade surgical instruments for spay procedures. This product contains: (1) No. 3 Scalpel Handle (1) Straight Mayo Scissor (1) Snook Ovariectomy Hook (2) Curved Kelly Forceps (1) Tissue Forceps with 1x2 Teeth (1) Allis Tissue Forceps with 4x5 Teeth (4) Backhous Towel Clamps (1) Brown-Adson Forceps with 7x7 Teeth (1) Olsen Hegar Needle Holder (2) Curved Halsted Mosquito Forceps (2) Curved Rochester-Carmalt Forceps (1) Sterilization/Storage Cassette
Catalog No. 50-283-1606 Supplier Med Vet International Supplier No. MILV96820
The Miltex Vantage V96820 feline spay pack contains 19 mid-grade surgical instruments for spay procedures. This product contains: (1) No. 3 Scalpel Handle (1) Straight Mayo Scissor (1) Snook Ovariectomy Hook (2) Curved Kelly Forceps (1) Tissue Forceps with 1x2 Teeth (1) Allis Tissue Forceps with 4x5 Teeth (4) Backhous Towel Clamps (1) Brown-Adson Forceps with 7x7 Teeth (1) Olsen Hegar Needle Holder (2) Curved Halsted Mosquito Forceps (2) Curved Rochester-Carmalt Forceps (1) Sterilization/Storage Cassette