Learn More
Med Vet International Miltex Vantage Veterinary Canine Spay Pack
Catalog No. 502829662
Encompass_Preferred
The Miltex Vantage V98610 canine spay pack contains 19 mid-grade surgical instruments for spay procedures. This product contains: (1) No. 3 Scalpel Handle (1) Straight Mayo Scissor (1) Snook Ovariectomy Hook (2) Curved Kelly Forceps (1) Tissue Forceps with 1x2 Teeth (1) Allis Tissue Forceps with 4x5 Teeth (4) Backhous Towel Clamps (1) Brown-Adson Forceps with 7x7 Teeth (1) Olsen Hegar Needle Holder (2) Curved Halsted Mosquito Forceps (2) Curved Rochester-Carmalt Forceps (1) Sterilization/Storage Cassette
Catalog No. 50-282-9662 Supplier Med Vet International Supplier No. MILV96810
The Miltex Vantage V98610 canine spay pack contains 19 mid-grade surgical instruments for spay procedures. This product contains: (1) No. 3 Scalpel Handle (1) Straight Mayo Scissor (1) Snook Ovariectomy Hook (2) Curved Kelly Forceps (1) Tissue Forceps with 1x2 Teeth (1) Allis Tissue Forceps with 4x5 Teeth (4) Backhous Towel Clamps (1) Brown-Adson Forceps with 7x7 Teeth (1) Olsen Hegar Needle Holder (2) Curved Halsted Mosquito Forceps (2) Curved Rochester-Carmalt Forceps (1) Sterilization/Storage Cassette