Learn More
Hillrom Veteriniary Ear Speculum
Catalog No. 22355155
Change view
Click to view available options
Size (Metric):
4 mm
Catalog No. Size (Metric)
22-355-155 4 mm
1 options
This item is not returnable. View return policy
Catalog No. 22-355-155 Supplier Hillrom Supplier No. WEL 22064
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specula for Veterinary Operating and Pneumatic Otoscopes. Material is lightweight, yet very durable.
  • The specula may be autoclaved, boiled, or sterilized in solution and can be attached easily
  • The material is lightweight but durable
  • For use with Veterinary Pneumatic and Operating Otoscopes
  • Style: Round Tip
  • Material: Plastic
  • Length: 4 mm
Specifications
Product Type Otoscope Specula
Size (Metric) 4 mm
For Use With (Equipment) Veterinary Otoscope