Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ VetMAX™-Gold SIV Detection Kit

Catalog No. 4415200
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-152-00 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-152-00 Supplier Applied Biosystems™ Supplier No. 4415200
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The USDA licensed, VetMAX™-Gold SIV Detection Kit is a robust PCR diagnostic kit, enabling early detection of swine influenza virus in pigs and helping to minimize the spread of the disease.

  • A robust PCR diagnostic kit that enables early detection of swine influenza virus in pigs and helps to minimize the spread of the disease
  • The first and only USDA-licensed real-time PCR test for the rapid detection of SIV RNA purified from porcine nasal swab samples
  • The VetMAX™-Gold SIV Detection assay is a single-tube (or well) real-time RT-PCR assay in which SIV and Xeno™ internal positive control RNA is reverse transcribed into cDNA, then amplified and detected in real time using fluorescent TaqMan™ probes
  • Delivers accurate results within 1.5 hours
  • Kit is not intended for the differentiation of SIV subtypes

Confidence in Results

  • The VetMAX™-Gold SIV Detection Kit utilizes AmpliTaq Gold™ DNA Polymerase and optimized primers and TaqMan™ probes for maximum sensitivity and specificity
  • Additionally, the included Xeno™ RNA internal positive control enables monitoring of RNA purification efficiency and PCR inhibition, allowing for increased accuracy
  • Designed to fit within a simple streamlined workflow, the VetMAX™-Gold SIV Detection Kit is optimized to perform with the MagMAX™ Pathogen RNA/DNA Kit and the Applied Biosystems™ 7500 Fast Real-Time PCR Systems
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -30°C to -10°C; Includes 100 reactions with the following components:

• Multiplex RT-PCR buffer
• Multiplex enzyme mix
• Nuclease-free water
• SIV primer-probe mix
• SIV-Xeno RNA control mix
• XenoRNA control
Detection Method Primer-Probe Detection
Format Kit
Sample Type Swab
For Use With (Application) Sequencing, Real-Time PCR
No. of Reactions 100
PCR Method 1-Step qRT-PCR
Product Line VetMAX
Product Type Gold SIV Detection Kit
Quantity 100 reactions
Shipping Condition Dry Ice
Species Swine/Porcine
Target Organism Class Swine Influenza Virus (SIV)
Show More Show Less
What are the quenchers in the VetMAX-Gold SIV Detection Kit?

The VetMAX-Gold SIV Detection Kit contains the following quenchers:
- SIV RNA: Eclipse® Q
- Xeno RNA Control: Eclipse® Q

What are the assay-labeled reporters in the VetMAX-Gold SIV Detection Kit?

The VetMAX-Gold SIV Detection Kit contains the following assay-labeled reporters:
- SIV RNA: FAM dye
- Xeno RNA Control: VIC dye

What is the internal positive control (IPC) in the VetMAX-Gold SIV Detection Kit?

The VetMAX-Gold SIV Detection Kit contains the Xeno RNA Control as the internal positive control (IPC) for the RNA purification process and to monitor for the presence of PCR inhibitors.

For Veterinary Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.