Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ 3-Methyl-2-quinoxalinecarboxylic acid, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015396 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5396 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5396 Supplier MilliporeSigma Supelco Supplier No. 3286250MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line VETRANAL
CAS 74003-63-7
Physical Form Neat
Linear Formula C10H8N2O2
MDL Number MFCD03840415
Type Analytical Standard
Quantity 50 mg
Formula Weight 188.18
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.