Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ 6-Methylprednisolone, MilliporeSigma™ Supelco™

Catalog No. 111015647 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5647 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5647 Supplier MilliporeSigma Supelco Supplier No. 46436100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 83-43-2
Shipping Condition Ambient
Physical Form Neat
Linear Formula C22H30O5
MDL Number MFCD00010591
Synonym 11 beta,17 alpha,21-Trihydroxy-6 alpha-methyl-1,4-pregnadiene-3,20-dione; 6 alpha-Methyl-11 beta,17 alpha,21-trihydroxy-1,4-pregnadiene-3,20-dione; Medrol; Medrone
Type Analytical Standard
Quantity 100 mg
Formula Weight 374.47
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.