Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Altrenogest, MilliporeSigma™ Supelco™

Catalog No. 111015682 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5682 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5682 Supplier MilliporeSigma Supelco Supplier No. 33994100MGR
Add to Cart
Add to Cart

Altrenogest can be called as a steroid or a progestational drug, which finds applications in breeding. It can mainly be used to control or suppress the condition of estrus in horses.

TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 850-52-2
Physical Form Neat
Linear Formula C21H26O2
MDL Number MFCD00867855
Synonym 17-alpha-Allyl-17-beta-hydroxyoestra-4,9,11-trien-3-one; Allyltrenbolone; Regumate
Type Analytical Standard
Quantity 100 mg
Formula Weight 310.43
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.