Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Ampicillin trihydrate, MilliporeSigma™ Supelco™

Catalog No. 111015337 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5337 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5337 Supplier MilliporeSigma Supelco Supplier No. 31591250MG
Add to Cart
Add to Cart

Chemical structure: ß-lactam

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 7177-48-2
Physical Form Neat
Linear Formula C16H19N3O4 S · 3H2 O
MDL Number MFCD00072036
Melting Point 198°C to 200°C
Synonym D-(-)-alpha-Aminobenzylpenicillin
Type Analytical Standard
Quantity 250 mg
Formula Weight 403.45
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.