Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Amprolium hydrochloride, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3021 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3021 Supplier MilliporeSigma Supelco Supplier No. 31592250MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 137-88-2
Physical Form Neat
Linear Formula C14H19N4 Cl · HCl
MDL Number MFCD00078831
Synonym 1-([4-Amino-2-propyl-5-pyrimidinyl]methyl)-2-methylpyridinium chloride
Type Analytical Standard
Quantity 250 mg
Formula Weight 315.24
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.