Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ AOZ, MilliporeSigma™ Supelco™

Catalog No. 111015598 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5598 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5598 Supplier MilliporeSigma Supelco Supplier No. 3334750MGR
Add to Cart
Add to Cart

3-Amino-2-oxazolidinone, a nitrofuran metabolite, which is bound to the tissues can be determined, using monoclonal enzyme-linked immunosorbent assay(ELISA)method.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 80-65-9
Physical Form Neat
Linear Formula C3H6N2O2
MDL Number MFCD00020871
Melting Point 65°C to 67°C
Synonym 3-Amino-2-oxazolidinone; NSC 111187; NSC 196570; NSC 38250
Type Analytical Standard
Quantity 50 mg
Formula Weight 102.09
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.