Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Arecoline hydrobromide, MilliporeSigma™ Supelco™

Catalog No. 111013850 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3850 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3850 Supplier MilliporeSigma Supelco Supplier No. 31593250MG
Add to Cart
Add to Cart

Arecoline hydrobromide is an arecoline salt of hydrobromic acid generally used in removing parasites from dog's intestine. It has a strong parasympathomimetic action.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 300-08-3
Physical Form Neat
Linear Formula C8H13 NO2 · HBr
MDL Number MFCD00039041
Melting Point 172°C to 176°C
Synonym 1-Methyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid methyl ester hydrobromide; Arecaidine methyl ester hydrobromide; Methyl 1,2,5,6-tetrahydro-1-methyl-3-pyridinecarboxylate hydrobromide; Methyl 1-methyl-1,2,5,6-tetrahydronicotinate hydrobromide; Taeniolin
Type Analytical Standard
Quantity 250 mg
Formula Weight 236.11
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.