Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Brombuterol hydrochloride, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013559 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3559 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3559 Supplier MilliporeSigma Supelco Supplier No. 9497210MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brombuterol hydrochloride is a beta-sympathomimetic compound with tocolytic, muscle-relaxant and bronchodilator properties and is widely used in therapy for their growth promoting and performance enhancement.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line VETRANAL
CAS 21912-49-2
Physical Form Neat
Linear Formula C12H18 Br2N2 O · HCl
MDL Number MFCD10567368
Synonym 1-(4-Amino-3,5-dibromophenyl)-2-tert-butylaminoethanol hydrochloride
Type Analytical Standard
Quantity 10 mg
Formula Weight 402.55
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.