Learn More
VETRANAL™ Canamycin A Disulfate Salt Hydrate Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 64013-70-3 (Anhydrous) |
| Physical Form | Neat |
| Molecular Formula | C18H36N4O11 · 2H2SO4 · xH2O |
| Linear Formula | C18H36N4O11 · 2H2SO4 · xH2O |
| UN Number | NONH for all modes of transport |
| Synonym | Kanamycin A disulfate salt |
| Formula Weight | 680.66 (Anhydrous Basis) |
| Grade | Analytical |
VETRANAL is a registered trademark of Sigma-Aldrich Chemie GmbH
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.