Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Carbuterol acetate, MilliporeSigma™ Supelco™

Catalog No. 111013286 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3286 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3286 Supplier MilliporeSigma Supelco Supplier No. 3238210MG
Add to Cart
Add to Cart

Specifications

Product Line VETRANAL
CAS 1613439-57-8
Linear Formula C13H21N3O3 · C2H4O2
MDL Number MFCD28118922
Type Analytical Standard
Quantity 10 mg
Formula Weight 327.38
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.