Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Cefquinome sulfate, MilliporeSigma™ Supelco™

Catalog No. 111012691 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2691 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2691 Supplier MilliporeSigma Supelco Supplier No. 32472100MG
Add to Cart
Add to Cart

Cefquinome sulfates are veterinary, parenteral, fourth-generation cephalosporins belonging to the aminothiazolyl class of compounds. They exhibit broad-spectrum antimicrobial activity against gram-positive and gram-negative bacteria such as Pseudomonas aeruginosa and Enterobacteriaceae.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 118443-89-3
Physical Form Neat
Linear Formula C23H24N6O5S2 · H2 SO4
MDL Number MFCD23381671
Synonym (6 R,7 R)-7-[[(2 Z)-2-(2-Amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-3-(5,6,7,8-tetrahydroquinolin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0 ]oct-2-ene-2-carboxylate sulfate; Cefquinome sulphate
Type Analytical Standard
Quantity 100 mg
Formula Weight 626.68
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.