Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Chloramphenicol, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014651 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4651 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4651 Supplier MilliporeSigma Supelco Supplier No. 31667250MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 56-75-7
Physical Form Neat
Linear Formula Cl2 CHCONHCH(CH2 OH)CH(OH)C6H4 NO2
MDL Number MFCD00078159
Melting Point 148°C to 150°C
Synonym D-(-)-threo-2,2-Dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-beta-(4-nitrophenyl)ethyl]acetamide; D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol; D-threo-2,2-Dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-4-nitrophenethyl]acetamide; Chloromycetin
Solubility Insoluble in water
Type Analytical Standard
Quantity 250 mg
Formula Weight 323.13
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.