Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Chlorpromazine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111019975 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9975 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9975 Supplier MilliporeSigma Supelco Supplier No. 31679250MG
Add to Cart
Add to Cart

Chlorpromazine hydrochloride (CPZ) is an antipsychotic drug which belongs to phenothiazine neuroleptic group. It is used in treating mental or personality disorders.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 69-09-0
Physical Form Neat
Linear Formula C17H19 ClN2 S · HCl
MDL Number MFCD00012654
Synonym 2-Chloro-10-(3-dimethylaminopropyl)phenothiazine hydrochloride; CPZ; Largactil
Type Analytical Standard
Quantity 250 mg
Formula Weight 355.33
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.