Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Cimbuterol, MilliporeSigma™ Supelco™

Catalog No. 111014550 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4550 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4550 Supplier MilliporeSigma Supelco Supplier No. 3257610MG
Add to Cart
Add to Cart

Cimbuterol is classified under the β-agonist group of compounds.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 54239-39-3
Physical Form Neat
Linear Formula C13H19N3 O
MDL Number MFCD16652572
Synonym 2-Amino-5-(2-tert-butylamino-1-hydroxyethyl)benzonitrile
Type Analytical Standard
Quantity 10 mg
Formula Weight 233.31
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.