Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Clorsulon, MilliporeSigma™ Supelco™

Catalog No. 111014844 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4844 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4844 Supplier MilliporeSigma Supelco Supplier No. 33973100MGR
Add to Cart
Add to Cart

Clorsulon, is a benzene sulphonamide and a flukicide, which is found to be effective against the adult and immature form of liver fluke, Fasciola hepatica in cattle.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 60200-06-8
Physical Form Neat
Linear Formula C8H8 Cl3N3O4S2
MDL Number MFCD00072006
Synonym 4-Amino-6-(trichlorovinyl)-1,3-benzenedisulfonamide
Type Analytical Standard
Quantity 100 mg
Formula Weight 380.66
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.