Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Crystal Violet, MilliporeSigma™ Supelco™

Catalog No. 111019417 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9417 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9417 Supplier MilliporeSigma Supelco Supplier No. 46364250MG
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 548-62-9
Physical Form Neat
Linear Formula C25H30N3 Cl
MDL Number MFCD00011750
Melting Point 205°C
Synonym Basic Violet 3; Gentian Violet; Hexamethylpararosaniline chloride; Methyl Violet 10 B
Solubility Soluble in water
Type Analytical Standard
Quantity 250 mg
Formula Weight 407.98
Grade Analytical Standard
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.