Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Cyclosporin A, MilliporeSigma™ Supelco™

Catalog No. 111014816 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4816 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4816 Supplier MilliporeSigma Supelco Supplier No. 32425100MG
Add to Cart
Add to Cart

Cyclosporin A has been used to:. Prevent rejection of kidney and liver transplants.. In graft versus host disease.. Treatment of autoimmune diseases. . Treatment of Schistosomiasis.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 59865-13-3
Physical Form Neat
Linear Formula C62H111N11O12
MDL Number MFCD00274558
Synonym Antibiotic S 7481F1; Cyclosporine
Type Analytical Standard
Quantity 100 mg
Formula Weight 1202.61
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.