Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Diethylstilbestrol, MilliporeSigma™ Supelco™

Catalog No. 111020200 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0200 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-0200 Supplier MilliporeSigma Supelco Supplier No. 46207250MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Diethylstilbestrol (DES) belongs to the synthetic oestrogens class of compounds.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 56-53-1
Physical Form Neat
Linear Formula C18H20O2
MDL Number MFCD00002373
Synonym (E)-3,4-Bis(4-hydroxyphenyl)-3-hexene; DES; Stilbestrol
Type Analytical Standard
Quantity 250 mg
Formula Weight 268.35
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.