Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Furaltadone, MilliporeSigma™ Supelco™

Catalog No. 111013046 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3046 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3046 Supplier MilliporeSigma Supelco Supplier No. 46289250MG
Add to Cart
Add to Cart

Furaltadone is a nitrofuran drug, with broad-spectrum antimicrobial activity. It is widely used in veterinary medicine for intensive animal production, especially in pigs, poultry, fish and to provide resistance against infections caused by Escherichia coli and Salmonella.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 139-91-3
Physical Form Neat
Linear Formula C13H16N4O6
MDL Number MFCD00057356
Synonym 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone
Type Analytical Standard
Quantity 250 mg
Formula Weight 324.29
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.