Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Griseofulvin-13C17 Solution, 25 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017930 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7930 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7930 Supplier MilliporeSigma Supelco Supplier No. 324911.2ML
Add to Cart
Add to Cart

Certan Vial

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 1325307-58-1
Flash Point 2.0°C - Closed Cup
Linear Formula 13C17H17 ClO6
MDL Number MFCD19704817
Synonym (2 S)-trans-7-Chloro-2 ′,4,6-trimethoxy-6 ′-methylspiro(benzofuran-2[3 H],1 ′-[2 ]cyclohexene)-3,4 ′-dione-13C17 solution
Type Analytical Standard
Quantity 1.2 mL
Formula Weight 369.64
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 25 μg/mL in acetonitrile
Show More Show Less

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.