Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ Halofuginone hydrobromide, MilliporeSigma™ Supelco™

Catalog No. 111018105 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8105 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8105 Supplier MilliporeSigma Supelco Supplier No. 3248110MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Halofuginone hydrobromide (Halofuginone) is a specific collagen Type I inhibitor that antagonize or inhibit the development of new blood vessels, hence can prevent intimal hyperplasia at a vascular anastomosis. It is used in the treatment or prevention of coccidiosis in both humans and animals.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line VETRANAL
CAS 64924-67-0
Physical Form Neat
Linear Formula C16H17 BrClN3O3 · HBr
MDL Number MFCD23843776
Synonym trans-(+/-)-7-Bromo-6-chloro-3-[3-(3-hydroxy-2-piperidinyl)-2-oxopropyl]-4(3 H)-quinazolinone monohydrobromide
Type Analytical Standard
Quantity 10 mg
Formula Weight 495.59
Grade Analytical Standard

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.